Learn More
Invitrogen™ Mouse anti-Human IgG Fc (CH2 domain) Secondary Antibody
Shop All Thermo Scientific ProductsDescription
This antibody recognizes human IgG Fc (all subclasses). The epitope detected lies within the CH2 domain as determined by haemagglutination and western blotting using IgG heavy chain and myelomas with defined domain deletions. The epitope is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. MA5-16859 cross-reacts with Rhesus monkey.
Specifications
Specifications
| Antigen | Human IgG Fc (CH2 domain) |
| Applications | ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | MK 1 A6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Host Species | Mouse |
| Immunogen | Human IgG Polyclonal. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.