Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG Fc Secondary Antibody (EM-07), Janelia Fluor 549, Novus Biologicals™
SDP

Catalog No. NB538934
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB538934 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB538934 Supplier R&D Systems Supplier No. NBP145063JF549

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgG Fc Monoclonal specifically detects IgG Fc in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Monoclonal
Clone EM-07
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Fusion protein of human IgG Fc fragment.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.