Learn More
Mouse anti-Human IgG Fc Secondary Antibody (MK 1 A6), DyLight 755, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | MK 1 A6 |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Human IgG Polyclonal |
| Purification Method | Protein A purified |
| Quantity | 0.2 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.