Learn More
Genscript Corporation Mouse Anti-Human IgG Fc Antibody (50B4A9)[HRP], mAb
Catalog No. 502271974
Encompass
GenScript Mouse Anti-Human IgG Fc Antibody (50B4A9)[HRP], mAb reacts with the Fc portion of human IgG but not with the Fab portion of human IgG. This monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken or goat immunoglobulins.
This item is not returnable. View return policy
Catalog No. 50-227-1974 Supplier Genscript Corporation Supplier No. A01854200
GenScript Mouse Anti-Human IgG Fc Antibody (50B4A9)[HRP], mAb reacts with the Fc portion of human IgG but not with the Fab portion of human IgG. This monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken or goat immunoglobulins.