Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Human IgG1, Fc Fragment Specific (HP6069), Clone: HP6069, Secondary Antibody,

Catalog No. 411451400UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
400 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
41-145-1400UG 400 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-145-1400UG Supplier MilliporeSigma™ Supplier No. 411451400UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects IgG1, Fc Fragment Specific (HP6069) Clone: HP6069 in Human samples, and it is validated for ELISA

The HP series of murine human Ig-specific monoclonal antibodies was produced from documented hybridoma cell lines that were developed at the Centers for Disease Control in Atlanta, Georgia. Laboratory analysis of each monoclonal antibody lot is performed in two stages during the production of ascites. First, the cell culture medium is analyzed for the presence and relative amount of human Ig-specific antibody by ELISA before injection of cells into mice. Second, collected ascites are analyzed for potency and antibody specificity using ELISA and isoelectric focusing (IEF), often in combination with an affinity immunoblot. For more information, CALBIOCHEM™ technical monograph, The Human IgG Subclasses, is available free of charge.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1, Fc Fragment Specific (HP6069)
Applications ELISA
Classification Monoclonal
Clone HP6069
Conjugate Unconjugated
Formulation In PBS, pH 7.4.
Host Species Mouse
Quantity 400 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.