Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG1 Fc Secondary Antibody (2C11cc), Biotin, Novus Biologicals™
SDP

Catalog No. NB459202
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB459202 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB459202 Supplier R&D Systems Supplier No. NB1002046B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgG1 Fc Monoclonal specifically detects IgG1 Fc in Human samples. It is validated for ELISA, Immunodiffusion, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen IgG1 Fc
Applications ELISA, Immunodiffusion, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2C11cc
Conjugate Biotin
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunodiffusion
Formulation PBS
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.