Learn More
Mouse anti-Human IgG2, Clone: G18-21, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The G18-21 antibody reacts specifically with human IgG2. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: G18-21
Isotype: IgG1, κ
Species Reactivity: Human
ELISA
Specifications
Specifications
| Antigen | Mouse anti-Human IgG2 |
| Classification | Monoclonal |
| Clone | G18-21 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.