Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Human IgG2, Fc Fragment Specific, Alkaline Phosphatase, Secondary Antibody,

Catalog No. 401462250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
Conjugate:
Alkaline Phosphatase (AP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-146-2250UG 250 μg Alkaline Phosphatase (AP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-146-2250UG Supplier MilliporeSigma™ Supplier No. 401462250UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects IgG2, Fc Fragment Specific Clone: in Human samples, and it is validated for ELISA, Enzyme Immunoassay

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2, Fc Fragment Specific
Applications ELISA
Classification Monoclonal
Conjugate Alkaline Phosphatase (AP)
Formulation In 50mM Tris-HCl, 1mM MgCl2, 50% glycerol, pH 8.0.
Host Species Mouse
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 2°C to 8°C
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.