Learn More
Mouse anti-Human IgG3 Heavy Chain Secondary Antibody (5G12cc), Janelia Fluor 549, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG3 Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 5G12cc |
| Conjugate | Janelia Fluor 549 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG3 from human serum. |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.