Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse anti-Human IgG3 (Heavy chain) Secondary Antibody, Biotin

Catalog No. 053640
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-364-0 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-364-0 Supplier Invitrogen™ Supplier No. 053640
  • $222.00
    $166.65 / Each of 1

    Save $55.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Mouse anti-Human IgG3 (Heavy chain) Secondary Antibody for ELISA

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG3 (Heavy chain)
Applications ELISA
Classification Monoclonal
Clone HP6047
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 40% glycerol, 1% BSA and 0.1% sodium azide; pH 7.4
Host Species Mouse
Immunogen Human IgG3.
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.