Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Human IgG3, Hinge Specific (HP6047), Biotin, Clone: HP6047, Secondary Antibody,

Catalog No. 411483200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
Conjugate:
Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
41-148-3200UG 200 μg Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-148-3200UG Supplier MilliporeSigma™ Supplier No. 411483200UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects IgG3, Hinge Specific (HP6047) Clone: HP6047 in Human samples, and it is validated for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG3, Hinge Specific (HP6047)
Applications ELISA
Classification Monoclonal
Clone HP6047
Conjugate Biotin
Formulation In PBS, 1% BSA, pH 7.4.
Host Species Mouse
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage −20°C
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.