Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG4, Clone: JDC-14, Secondary Antibody, BD

Catalog No. BDB555878 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555878 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555878 Supplier BD Biosciences Supplier No. 555878
Add to Cart
Add to Cart

The JDC-14 antibody reacts specifically with human IgG4. It does not react with other Ig isotypes.

Host Species: Mouse
Clone: JDC-14
Isotype: IgG1, κ
Species Reactivity: Human

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Human IgG4
Classification Monoclonal
Clone JDC-14
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Human
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.