Learn More
Immunology Consultants anti-Human IgM Antibody (Mouse Clone #208)
SDP
Catalog No. 502248899
Encompass_Preferred
Clone 208 targets a defined epitope on human IgM, providing complementary detection to the standard MM‑80A reagent. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal background. MM‑80A‑208 is well suited for immunology research, diagnostic assay development, and studies examining early immune responses. Its consistent lot‑to‑lot performance ensures reliable IgM detection across diverse sample matrices.
This item is not returnable. View return policy
Catalog No. 50-224-8899 Supplier Immunology Consultants Supplier No. MM80A208
Clone 208 targets a defined epitope on human IgM, providing complementary detection to the standard MM‑80A reagent. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal background. MM‑80A‑208 is well suited for immunology research, diagnostic assay development, and studies examining early immune responses. Its consistent lot‑to‑lot performance ensures reliable IgM detection across diverse sample matrices.