Learn More
Immunology Consultants anti-Human IgM Antibody (Mouse) - Monoclonal
SDP
Catalog No. 502248898
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of human IgM, supporting ELISA, Western blotting, and immunoassay development. It delivers strong affinity and low nonspecific binding, enabling precise measurement of IgM in serum, plasma, and purified immunoglobulin preparations. MM‑80A is ideal for infectious disease research, immune profiling, and diagnostic assay development requiring reliable IgM quantification. Its clean, reproducible signal ensures dependable performance across repeated assays and varied sample types.
Catalog No. 50-224-8898 Supplier Immunology Consultants Supplier No. MM80A
This monoclonal antibody provides sensitive, specific detection of human IgM, supporting ELISA, Western blotting, and immunoassay development. It delivers strong affinity and low nonspecific binding, enabling precise measurement of IgM in serum, plasma, and purified immunoglobulin preparations. MM‑80A is ideal for infectious disease research, immune profiling, and diagnostic assay development requiring reliable IgM quantification. Its clean, reproducible signal ensures dependable performance across repeated assays and varied sample types.