Learn More
Mouse anti-Human IgM, Clone: G20-127, PE, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The G20-127 antibody reacts specifically with the heavy chain of human IgM. It does not react with other heavy chain isotypes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G20-127
Isotype: IgG1, κ
Species Reactivity: Human
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Human IgM |
| Classification | Monoclonal |
| Clone | G20-127 |
| Conjugate | PE |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.