Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse anti-Human IgM Fc Secondary Antibody

Catalog No. 01676767 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-767 Supplier Invitrogen™ Supplier No. SA119256
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

This antibody reacts with Fc fragment of human IgM.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgM Fc
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CH2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Host Species Mouse
Immunogen Purified human IgM.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.