Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse anti-Human Kappa Light Chain Secondary Antibody, Biotin

Catalog No. PIMA512114
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512114 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512114 Supplier Invitrogen™ Supplier No. MA512114
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Secondary Antibody

By Western blot, MA5-12114 detects Human Ig kappa light chain chain at ~25kD in human samples. At high concentrations, the antibody will also detect heavy chain. By ELISA and WB, MA5-12114 does not react with human Ig lambda light chain.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Kappa Light Chain
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Flow Cytometry, ELISA
Classification Monoclonal
Clone L1C1
Concentration 0.2 mg/mL
Conjugate Biotin
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Alias Immunoglobulin kappa light chain, Ig kappa light chain, Ig kappa, kappa light chain, Igk
Host Species Mouse
Immunogen B lymphoma cells.
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.