Learn More
Invitrogen™ Mouse anti-Human Lambda Light Chain Secondary Antibody, PE
Shop All Thermo Scientific ProductsDescription
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human Lambda Light Chain antibody, clone Mc24-IC6 recognizes human lambda light chains, and shows no cross- reactivity to kappa light chains or to immunoglobulin heavy chains.
Specifications
Specifications
| Antigen | Human Lambda Light Chain |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | Mc24-IC6 |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 5% sucrose, 1% BSA and 0.09% sodium azide; pH 7.4 |
| Host Species | Mouse |
| Immunogen | Purified human IgG. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.