Learn More
Chondrex Inc MOUSE ANTI-LPS IGG ASSAY KT
Catalog No. 501526792
Encompass_Preferred
Assay kit to determine mouse anti-lipopolysaccharide (LPS) IgG antibodies. Kit comes with 96-well ELISA plate pre-coated with LPS (from E. coli O111:B4) and all reagents necessary to complete the assay. Standard Range: 100 ng/mL to 1.6 ng/mL. Working Time: 4.5 Hours.
Catalog No. 50-152-6792 Supplier Chondrex Inc Supplier No. 6106
Assay kit to determine mouse anti-lipopolysaccharide (LPS) IgG antibodies. Kit comes with 96-well ELISA plate pre-coated with LPS (from E. coli O111:B4) and all reagents necessary to complete the assay. Standard Range: 100 ng/mL to 1.6 ng/mL. Working Time: 4.5 Hours.