Learn More
Biolegend Purified anti-mouse Ly-6C (Maxpar Ready)
Supplier:  Biolegend 128039
Encompass_Preferred
Purified anti-mouse Ly-6C (Maxpar Ready) [HK1.4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1365729