Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Anti-Monkey IgG Antibody [5D12A12] (biotin), ProSci
SDP

Catalog No. 16916855 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-916-855 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-916-855 Supplier ProSci LLC Supplier No. 16060100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Secondary Antibody

Mouse Anti-Monkey IgG Antibody specifically detects Mouse Anti-Monkey IgG in Monkey Samples. It is validated for ELISA, Western Blot, Immunohistochemistry.

Specifications

Antigen Mouse Anti-Monkey IgG
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 5D12A12
Concentration 1 mg/mL
Conjugate Biotin
Dilution Immunoblotting: 1/1,000 - 1/20,000, ELISA: 1/1,000 - 1/20,000, Immunohistochemistry: 1/100 - 1/1,000
Formulation PBS pH 7.2, 1% BSA, 0.02% Sodium Azide
Host Species Mouse
Immunogen Monkey IgG
Purification Method The purified antibody-Biotin conjugate is separated from non-conjugated biotin by dialysis.
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No cross reactivity with immunoglobulins from other species.
Target Species Monkey
Content And Storage Store at 2-8°C. The product is stable for about 4-6 weeks at 4°C as an undiluted liquid. For extended storage, aliquot and freeze the product at -70°C or below. Avoid repeated freezing and thawing. Or alternatively, add an equal volume of glycerol for a final concentration of 50% and store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.