Learn More
IgG Mouse anti-Rhesus Ma, Cynomolgus, Unconjugated, Clone: SB108a, Southern Biotech™
Shop All Southern Biotechnology Associates Products

Description
SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
Specifications
| Antigen | IgG |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | SB108a |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Formulation | Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added |
| Host Species | Mouse |
| Immunogen | Rhesus IgG |
| Quantity | 0.5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.