Learn More
Mouse anti-Mouse IgD[b], Clone: 217-170, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The 217-170 antibody reacts specifically with mouse IgD of Igh-C[b] haplotype (eg, C57BL/6, C57BL/10, SJL). It does not react with IgD of Igh-C[a] or related haplotypes (eg, BALB/c, AKR, CBA, C3H/He, C58, DBA/1, DBA/2, NZB, PL, RIII), or Igh-C[e] haplotyp
Host Species: Mouse
Clone: 217-170
Isotype: IgG1, κ
Species Reactivity: Mouse
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Mouse IgD[b] |
| Classification | Monoclonal |
| Clone | 217-170 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.