Learn More
Mouse anti-Mouse IgG2a[a], Clone: 8.3, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The 8.3 antibody reacts specifically with mouse IgG2a of Igh-Ca and the d, e, f, g, h, j, n, and o haplotypes (eg, BALB/c, C58, and A, AKR, CBA, C3H/He, NZB). It does not react with IgG2a of Igh-Cb nor the c and p haplotypes (eg, C57BL/6, C57BL/10, SJL, D
Host Species: Mouse
Clone: 8.3
Isotype: IgG2a, κ
Species Reactivity: Mouse
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Mouse IgG2a[a] |
| Classification | Monoclonal |
| Clone | 8.3 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.