Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Mouse IgM[a], Clone: DS-1, Secondary Antibody, BD

Catalog No. BDB553514 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553514 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553514 Supplier BD Biosciences Supplier No. 553514
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DS-1 antibody reacts specifically with mouse IgM of Igh-C[a] and related haplotypes (e.g., BALB/c, C58, CBA, C3H/Bi, C3H/He, DBA/1, DBA/2). It does not react with IgM of Igh-C[b], Igh-C[e], or related hapltypes (e.g., C57BL, SJL, A, AKR, NZB). DS-1 an

Host Species: Mouse
Clone: DS-1
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Mouse NMRI Monoclonal Antibody TC31

ELISA, Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Mouse IgM[a]
Classification Monoclonal
Clone DS-1
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.