Learn More
Mouse anti-Mouse IgM[b], Clone: AF6-78, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The AF6-78 antibody reacts specifically with mouse IgM of Igh-C[b] and related haplotypes (e.g., C57BL/6, C57BL/10, SJL, AKR, NZB). It does not react with IgM of Igh-C[a] or related haplotypes (e.g., BALB/c, C58, CBA, C3H/Bi, C3H/He, DBA/1, DBA/2, PL, RII
Host Species: Mouse
Clone: AF6-78
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: C57BL/10 mouse splenocytes
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Mouse IgM[b] |
| Classification | Monoclonal |
| Clone | AF6-78 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.