Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Mouse IgM, Secondary Antibody, BD

Catalog No. BDB562565 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562565 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562565 Supplier BD Biosciences Supplier No. 562565
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R6-60.2 antibody monoclonal antibody specifically binds to mouse Immunoglobulin M (IgM) of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. The R6-60.2 antibody has not been shown to stimulate B-cell proliferation. This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488nm), green (532nm) and yellow-green (561nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20nm filter).

Host Species: Rat
Clone: R6-60.2
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgM
Classification Monoclonal
Clone R6-60.2
Concentration 0.2mg/mL
Conjugate PE-CF594
Host Species Rat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.