Learn More
Alpha Diagnostic International Mouse Anti-nRNP Ig's ELISA Kit, Quantitative Immunoassay, Detects Total IgG, IgA, IgM in Mouse Samples
SDP
Supplier:  Alpha Diagnostic International 5410
Encompass
The Mouse Anti-nRNP Ig's ELISA Kit is a quantitative assay designed for detecting autoantibodies to native ribonuclear protein in mouse serum and plasma. This kit offers a highly sensitive method vital for researchers investigating autoimmune diseases like systemic lupus erythematosus. Unlike other kits, it ensures specificity by omitting detectable SS-A, SS-B, Scl-70, and Jo-1 antigens, utilizing a solid-phase immunoassay format for accurate results.
Catalog No. NC1180040