Learn More
Chondrex Inc MOUSE ANTI-OVA IGE E-C1, 1 MG
Supplier:  Chondrex Inc 3006
Encompass_Preferred
1 mg (lyophilized) or mouse anti-ovalbumin (OVA) monoclonal IgE antibody (Clone E-C1) that can be used to induce in vitro mast cell activation and in vivo hypersensitivity reactions. Can also be used for ELISA and Western blots. Special precautions must be taken when reconstituting the antibody, please contact Chondrex, Inc. at support@chondrex.com for more information.
Catalog No. 50-152-6735