Learn More
Chondrex Inc MOUSE ANTI-OVA IGM ASSAY KT
Supplier:  Chondrex Inc 3017
Encompass_Preferred
ELISA kit for assaying mouse anti-ovalbumin (OVA) IgM antibodies found in mouse serum and plasma samples. This kit includes one 96-well ELISA plate with removable strips, as well as all reagents necessary to complete the assay. This kit is ideal for quantifying the immune response to ovalbumin used to induce allergy models in mice. Standard Range: 12.5 ng/ml - 0.2 ng/ml. Working Time: 4.5 hours.
Catalog No. 50-152-6746