Learn More
Chondrex Inc Mouse Anti-Ovalbumin IgG1 Monoclonal Antibody, Clone 2322
Supplier:  Chondrex Inc 7094
Encompass_Preferred
Mouse Anti-Ovalbumin (OVA) IgG1 Monoclonal Antibody, Clone 2322, unconjugated Epitope: OVA 61-68: DKLPGFGD. Use for ELISA. Reacts to native OVA, denatured OVA, and OVA peptides. Purified by Protein G chromatography. Dilute the antibody in buffer containing 2% heterologous serum, such as normal goat serum, to prevent non-specific reactions in ELISA.
Catalog No. NC1929638