Learn More
Biotang Inc Cabozantinib,XL184, Free Base, > 99%, M.W. 501.51, C28H24FN3O5, CAS#849217-68-1. RS331-10MG
Catalog No. 50151460 Shop All BIOTANG Inc Products
Encompass_Preferred
Cabozantinib,XL184, Free Base, > 99%, M.W. 501.51, C28H24FN3O5, CAS#849217-68-1. RS331-10MG
Catalog No. 50-151-460 Supplier Biotang Inc Supplier No. BTANT228
Cabozantinib,XL184, Free Base, > 99%, M.W. 501.51, C28H24FN3O5, CAS#849217-68-1. RS331-10MG