Learn More
Santa Cruz Biotechnology MOUSE ANTI PCNA
Supplier:  Santa Cruz Biotechnology SC56
Encompass
PCNA (PC10),mouse monoclonal IgG2a; 200 microgram/ml,recommended for detection of PCNA p36 protein expressed at high levels in proliferating cells of m, r, h, insect and Schizosaccharomyces pombe origin by WB, IP, IHC(P) and FCM
Catalog No. NC9414864