Learn More
Life Diagnostics Inc Mouse Anti-PEG IgM ELISA Kit, Serum, Plasma, Detection Wavelength 450 nm, Standard Range Up to 100 u/ml, Kit Stability 6 months
SDP
Supplier:  Life Diagnostics Inc PEGM1N
Encompass_Preferred
Mouse Anti-PEG IgM ELISA Kit, Serum, Plasma, Detection Wavelength 450 nm, Standard Range Up to 100 u/ml, Kit Stability 6 months
Catalog No. NC1149468