Learn More
Genscript Corporation Mouse Anti-Rabbit IgG Fc Antibody (14H9H10)[HRP], mAb
Catalog No. NC2179119
Encompass
GenScript Mouse Anti-Rabbit IgG Fc Antibody (14H9H10)[HRP], mAb reacts with the Fc portion of rabbit IgG but not with the Fab portion of rabbit IgG. The monoclonal antibody has no cross-reactivity with human, mouse, rat, sheep, bovine, goat or camel immunoglobulins.
This item is not returnable. View return policy
Catalog No. NC2179119 Supplier Genscript Corporation Supplier No. A01856
GenScript Mouse Anti-Rabbit IgG Fc Antibody (14H9H10)[HRP], mAb reacts with the Fc portion of rabbit IgG but not with the Fab portion of rabbit IgG. The monoclonal antibody has no cross-reactivity with human, mouse, rat, sheep, bovine, goat or camel immunoglobulins.