Learn More
Genscript Corporation Mouse Anti-Rabbit IgG Fc Antibody (14H9H10)[HRP], mAb
Supplier:  Genscript Corporation A01856200
Encompass
GenScript Mouse Anti-Rabbit IgG Fc Antibody (14H9H10)[HRP], mAb reacts with the Fc portion of rabbit IgG but not with the Fab portion of rabbit IgG. The monoclonal antibody has no cross-reactivity with human, mouse, rat, sheep, bovine, goat or camel immunoglobulins.
Catalog No. 50-227-1976