Learn More
Santa Cruz Biotechnology MOUSE ANTI-RABBIT IGG PE-CY7,
Catalog No. NC1569698
Encompass
mouse anti-rabbit IgG PE-Cy7, 200 µg/0.5 ml•mouse monoclonal secondary antibody conjugated to phycoerythrin with cyanin-7 (PE-Cy7)•supplied at 200 µg in 0.5 ml volume•Strongly recommended alternative to polyclonal anti-rabbit IgG secondary antibodies for Immunofluorescence (IF), Immunohistochemistry (IHC) and Flow Cytometry (FCM)•Highly specific secondary antibody that provides strong signal with minimal background and virtually complete elimination of lot to lot variation associated with polyclonal secondary antibodies
Catalog No. NC1569698 Supplier Santa Cruz Biotechnology Supplier No. SC516721
mouse anti-rabbit IgG PE-Cy7, 200 µg/0.5 ml•mouse monoclonal secondary antibody conjugated to phycoerythrin with cyanin-7 (PE-Cy7)•supplied at 200 µg in 0.5 ml volume•Strongly recommended alternative to polyclonal anti-rabbit IgG secondary antibodies for Immunofluorescence (IF), Immunohistochemistry (IHC) and Flow Cytometry (FCM)•Highly specific secondary antibody that provides strong signal with minimal background and virtually complete elimination of lot to lot variation associated with polyclonal secondary antibodies