Learn More
Genscript Corporation Mouse Anti-Rabbit IgG Antibody (M205) [HRP], mAb
Catalog No. 502271967
Encompass
GenScript Anti-Rabbit IgG Antibody (M205) only reacts with native rabbit IgG and does not bind to the denatured or reduced rabbit IgG heavy chain or light chain. The monoclonal antibody has no cross-reactivity with mouse, rat, bovine, goat, or sheep immunoglobulins and limited cross-reactivity with human immunoglobulins.
This item is not returnable. View return policy
Catalog No. 50-227-1967 Supplier Genscript Corporation Supplier No. A01827200
GenScript Anti-Rabbit IgG Antibody (M205) only reacts with native rabbit IgG and does not bind to the denatured or reduced rabbit IgG heavy chain or light chain. The monoclonal antibody has no cross-reactivity with mouse, rat, bovine, goat, or sheep immunoglobulins and limited cross-reactivity with human immunoglobulins.