Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Anti-Rabbit IgM, Clone: 367.2, Secondary Antibody, BD

Catalog No. BDB550938 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550938 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550938 Supplier BD Biosciences Supplier No. 550938
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 367.2 antibody reacts specifically with rabbit IgM. It does not react with rabbit IgG or IgA isotypes.

Host Species: Mouse
Clone: 367.2
Isotype: IgG1 κ
Species Reactivity: Rabbit
Immunogen: Purified rabbit IgM

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse Anti-Rabbit IgM
Classification Monoclonal
Clone 367.2
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Mouse
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rabbit
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.