Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light chain Mouse anti-Rabbit, HRP , Secondary Antibody, MilliporeSigma™

Catalog No. MAB201PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB201PMI 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB201PMI Supplier MilliporeSigma Supplier No. MAB201P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Detect Rabbit light chain using this Mouse anti-Rabbit light chain, HRP conjugate Antibody validated for use in ELISA, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rabbit light chain
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate HRP
Formulation Purified from antiserum by chromatographic procedures. Lyophilized in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6. 15mg/mL BSA as stabilizer.
Host Species Mouse
Immunogen Prepared from rabbit IgG light chain.
Purification Method Serum
Quantity 0.5 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Keep away from light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.