Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat and anti-Hamster Ig, κ Light Chain, Clone: RG7/7.6, Secondary Antibody, BD

Catalog No. BDB550336 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550336 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550336 Supplier BD Biosciences Supplier No. 550336
Add to Cart
Add to Cart

The RG7/7.6 antibody reacts strongly with rat Igs bearing - light chain of the Igk-1[a]allotype (e.g., AO, AUG, BN, BUF, LEW, LOU, PVG, WAG, WF), weakly with Igk-1[b] light chains (e.g., ACI, COP, DA, F344, SHR), and it does not react with λ light chain nor heavy chains. At the time that this antibody was characterized, the Igk-1[a] allotype was designated - 1b, and Igk-1[b] was -1a; the current nomenclature was standardized at the Fourth International Workshop on Allogenic Systems in the Rat. The RG7/7.6 antibody also strongly cross-reacts with
Syrian and Armenian hamster Ig - light chain. This antibody is routinely tested by immunohistochemical analysis. Other applications were tested at BD Biosciences Pharmingen during
antibody development only or reported in the literature.

Host Species: Mouse
Clone: RG7/7.6
Isotype: IgG2a κ
Species Reactivity: Rat
Immunogen: Pooled Rat IgG

Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat and anti-Hamster Ig, κ Light Chain
Classification Monoclonal
Clone RG7/7.6
Conjugate Biotin
Host Species Mouse
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Hamster, Rat
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.