Learn More
Mouse anti-Rat Ig κ Light Chain, Clone: MRK-1, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The MRK-1 antibody reacts specifically with rat Igs bearing κ light chain. It does not react withλ light chain or heavy chains. MRK-1 antibody is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has not been shown to stimulate B-cell proliferation.
Host Species: Mouse
Clone: MRK-1
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Pooled rat Ig
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Mouse anti-Rat Ig κ Light Chain |
| Classification | Monoclonal |
| Clone | MRK-1 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.