Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat Ig κ light chain, Clone: RG7/9.1, Secondary Antibody, BD

Catalog No. BDB553901 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553901 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553901 Supplier BD Biosciences Supplier No. 553901

missing translation for 'validMainframeMsg'

The RG7/9.1 antibody reacts specifically with rat Igs bearing κ light chain and with free rat κ light chains. It does not react with λ light chain or heavy chains. The antibody cross-reacts with with Syrian hamster Ig, κ light chain. RG7/9.1 antibody has not been shown to stimulate B-cell proliferation.

Host Species: Mouse
Clone: RG7/9.1
Isotype: IgG2b
Species Reactivity: Rat
Immunogen: Pooled rat IgG

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat Ig κ light chain
Classification Monoclonal
Clone RG7/9.1
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rat
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.