Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgE Heavy Chain Secondary Antibody (MARE-1), Novus Biologicals™
SDP

Catalog No. NB002830
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002830 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002830 Supplier R&D Systems Supplier No. NBP268394250UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgE Monoclonal specifically detects IgE in Rat samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgE
Applications ELISA, Western Blot
Classification Monoclonal
Clone MARE-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias IgE, immunoglobulin heavy constant epsilon
Gene Symbols IGHE
Host Species Mouse
Immunogen Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins.
Purification Method Affinity Purified
Quantity 250 μg
Research Discipline Adaptive Immunity, Immunology
Gene ID (Entrez) 3497
Test Specificity Mouse anti Rat IgE antibody, clone MARE-1, recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin. The antibody binds to rat IgE with an avidity of 4x10^9 M^-1
Target Species Rat
Content And Storage Store at 4°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.