Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse anti-Rat IgE Secondary Antibody, FITC

Catalog No. SA125261 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
125 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA125261 125 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA125261 Supplier Invitrogen™ Supplier No. SA125261
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

SA1-25261 detects rat IgE. SA1-25261 is specific for the epsilon chain of rat immunoglobulin IgE and does not cross react with other classs of rat immunoglobulin. Use 50 μl of the suggested working dilution to label 2x10^7 cells in 100 μL.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgE
Applications Flow Cytometry
Classification Monoclonal
Clone MARE-1
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 50% glycerol and 0.1% sodium azide
Host Species Mouse
Immunogen Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins.
Purification Method Antigen affinity chromatography
Quantity 125 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.