Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The RG11/39.4 antibody reacts specifically with the Fc region of rat IgG1. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: RG11/39.4
Isotype: IgG2a, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Mouse anti-Rat IgG1 |
| Classification | Monoclonal |
| Clone | RG11/39.4 |
| Concentration | 0.5mg/mL |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?