Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 γ1 chainMouse anti-Rat, Alkaline Phosphatase , Secondary Antibody, MilliporeSigma™

Catalog No. MAB503AMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB503AMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB503AMI Supplier MilliporeSigma Supplier No. MAB503A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This Mouse anti-Rat IgG1 γ1 chain Antibody, Alkaline Phosphatase conjugate is validated for use in ELISA for the detection of Rat IgG1 γ1 chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat IgG1 γ1 chain
Applications ELISA
Classification Monoclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Purified by affinity chromatography. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.
Host Species Mouse
Immunogen Prepared from rat IgG1 γ1 chain.
Purification Method Affinity chromatography
Quantity 1 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.