Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 γ1 chainMouse anti-Rat, Cy5 , Secondary Antibody, MilliporeSigma™

Catalog No. MAB503SMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB503SMI 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB503SMI Supplier MilliporeSigma Supplier No. MAB503S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Mouse anti-Rat IgG1 γ1 chain Antibody, Cy5 conjugate is an antibody against Rat IgG1 γ1 chain for use in ELISA, immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat IgG1 γ1 chain
Applications ELISA, Immunofluorescence
Classification Monoclonal
Conjugate Cyanine5
Formulation Purified by affinity chromatography. Liquid in 1.0 mL of PBS/NaN3.
Host Species Mouse
Immunogen Prepared from rat IgG1 γ1 chain.
Purification Method Affinity chromatography
Quantity 0.1 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Do not Freeze.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.