Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgG2b, Clone: RG7/11.1, FITC, Secondary Antibody, BD

Catalog No. BDB553900 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553900 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553900 Supplier BD Biosciences Supplier No. 553900
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The RG7/11.1 antibody reacts specifically with the Fc region of rat IgG2b. It does not react with other Ig isotypes.

Host Species: Mouse
Clone: RG7/11.1
Isotype: IgG2b, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat IgG2b
Classification Monoclonal
Clone RG7/11.1
Concentration 0.5mg/mL
Conjugate FITC
Host Species Mouse
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rat
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.