Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Rat IgG2c gamma2c chain, Cy5, Secondary Antibody,

Catalog No. mab506smi Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.1 mg
Conjugate:
Cyanine5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB506SMI 0.1 mg Cyanine5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB506SMI Supplier MilliporeSigma™ Supplier No. MAB506S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects IgG2c γ2c chain Clone: in Rat samples, and it is validated for ELISA, Immunofluorescence

IgG is one of the most abundant protein in human serum. IgG is important for our defence against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2c γ2c chain
Applications ELISA, Immunofluorescence
Classification Monoclonal
Conjugate Cyanine5
Formulation Purified by affinity chromatography. Liquid in 1.0 mL of PBS/NaN3.
Host Species Mouse
Immunogen Prepared from rat IgG2c γ2c chain.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Secondary, Control Antibodies
Primary or Secondary Secondary
Test Specificity The antibody reacts with the γ2c heavy chain (Fc) of rat IgG2c.
Target Species Rat
Content And Storage 2°C to 8°C under sterile conditions for up to twelve months from date of receipt; Do not freeze
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.